Learn More
Abnova™ Human NDRG4 Full-length ORF (AAH11795.1, 1 a.a. - 339 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065009-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signalling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MPECWDGEHDIETPYGLLHVVIRGSPKGNRPAILTYHDVGLNHKLCFNTFFNFEDMQEITKHFVVCHVDAPGQQVGASQFPQGYQFPSMEQLAAMLPSVVQHFGFKYVIGIGVGAGAYVLAKFALIFPDLVEGLVLVNIDPNGKGWIDWAATKLSGLTSTLPDTVLSHLFSQEELVNNTELVQSYRQQIGNVVNQANLQLFWNMYNSRRDLDINRPGTVPNAKTLRCPVMLVVGDNAPAEDGVVECNSKLDPTTTTFLKMADSGGLPQVTQPGKLTEAFKYFLQGMGYMPSASMTRLARSRTASLTSASSVDGSRPQACTHSESSEGLGQVNHTMEVSCSpecifications
AAH11795.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.03kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686I1615/FLJ30586/FLJ42011/KIAA1180/MGC19632/SMAP-8 | |
NDRG4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
65009 | |
NDRG4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPECWDGEHDIETPYGLLHVVIRGSPKGNRPAILTYHDVGLNHKLCFNTFFNFEDMQEITKHFVVCHVDAPGQQVGASQFPQGYQFPSMEQLAAMLPSVVQHFGFKYVIGIGVGAGAYVLAKFALIFPDLVEGLVLVNIDPNGKGWIDWAATKLSGLTSTLPDTVLSHLFSQEELVNNTELVQSYRQQIGNVVNQANLQLFWNMYNSRRDLDINRPGTVPNAKTLRCPVMLVVGDNAPAEDGVVECNSKLDPTTTTFLKMADSGGLPQVTQPGKLTEAFKYFLQGMGYMPSASMTRLARSRTASLTSASSVDGSRPQACTHSESSEGLGQVNHTMEVSC | |
RUO | |
NDRG4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |