missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NCAPH2 Full-length ORF (NP_689512.1, 1 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_689512.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 29781 |
Molecular Weight (g/mol) | 88.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139102
|
Abnova™
H00029781-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129102
|
Abnova™
H00029781-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPH2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM]
Sequence: MNFIEAALLIQGSACVYSKKVEYLYSLVYQALDFISGKRRAKQLSSVQEDRANGVASSGVPQEAENEFLSLDDFPDSRTNVDLKNDQTPSEVLIIPLLPMALVAPDEMEKNNNPLYSRQGEVLASRKDFRMNTCVPHPRGAFMLEPEGMSPMEPAGVSPMPGTQKDTGRTEEQPMEVSVCRSPVPALGFSQEPGPSPEGPMPLGGGEDEDAEEAVELPEASAPKAALEPKESRSPQQSAALPRRYMLREREGAPEPASCVKETPDPWQSLDPFDSLESKPFKKGRPYSVPPCVEEALGQKRKRKGAAKLQDFHQWYLAAYADHADSRRLRRKGPSFADMEVLYWTHVKEQLETLRKLQRREVAEQWLRPAEEDHLEDSLEDLGAADDFLEPEEYMEPEGADPREAADLDAVPMSLSYEELVRRNVELFIATSQKFVQETELSQRIRDWEDTVQPLLQEQEQHVPFDIHTYGDQLVSRFPQLNEWCPFAELVAGQPAFEVCRSMLASLQLANDYTVEITQQPGLEMAVDTMSLRLLTHQRAHKRFQTYAAPSMAQPSpecifications
NP_689512.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNFIEAALLIQGSACVYSKKVEYLYSLVYQALDFISGKRRAKQLSSVQEDRANGVASSGVPQEAENEFLSLDDFPDSRTNVDLKNDQTPSEVLIIPLLPMALVAPDEMEKNNNPLYSRQGEVLASRKDFRMNTCVPHPRGAFMLEPEGMSPMEPAGVSPMPGTQKDTGRTEEQPMEVSVCRSPVPALGFSQEPGPSPEGPMPLGGGEDEDAEEAVELPEASAPKAALEPKESRSPQQSAALPRRYMLREREGAPEPASCVKETPDPWQSLDPFDSLESKPFKKGRPYSVPPCVEEALGQKRKRKGAAKLQDFHQWYLAAYADHADSRRLRRKGPSFADMEVLYWTHVKEQLETLRKLQRREVAEQWLRPAEEDHLEDSLEDLGAADDFLEPEEYMEPEGADPREAADLDAVPMSLSYEELVRRNVELFIATSQKFVQETELSQRIRDWEDTVQPLLQEQEQHVPFDIHTYGDQLVSRFPQLNEWCPFAELVAGQPAFEVCRSMLASLQLANDYTVEITQQPGLEMAVDTMSLRLLTHQRAHKRFQTYAAPSMAQP | |
RUO | |
NCAPH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
29781 | |
NCAPH2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
384D8-2/384D8_6/CAP-H2/MGC15858/MGC18000/MGC2455/MGC4133/MGC5305/MGC8640/hCAP-H2 | |
NCAPH2 | |
Recombinant | |
wheat germ expression system |