missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NBPF15 (aa 373-397) Control Fragment Recombinant Protein

Product Code. 30211102
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30211102 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30211102 Supplier Invitrogen™ Supplier No. RP108172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83644 (PA5-83644. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NBPF15 (NBPF Member 15), located on 1q21.1, is a Protein Coding gene. This gene is a member of the neuroblastoma breakpoint family (NBPF) which consists of dozens of recently duplicated genes primarily located in segmental duplications on human chromosome 1. This gene family contains numerous pseudogenes. Human NBPF15 encodes a 670 AA protein consisting of six clades of Olduvai domains. The entanglements between the mosaic disorder-ordered segments in the NBPF15 N terminus have triggered the multiple types of aggregates and phase transition of NBPF15 proteins, which could be associated with Olduvai-related cognitive dysfunction diseases. Diseases associated with NBPF15 include Neuroblastoma and Hydrolethalus Syndrome 1. An important paralog of this gene is NBPF14.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N660
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 284565
Name Human NBPF15 (aa 373-397) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AB14; NBPF; NBPF Member 15; Neuroblastoma Breakpoint Family, Member 1; Neuroblastoma Breakpoint Family, Member 16
Common Name NBPF15
Gene Symbol NBPF15
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SGCLELTDSCQPYRSAFYVLEQQRV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.