missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NAAA Partial ORF (NP_055250, 36 a.a. - 104 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027163-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138632

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

This gene encodes an N-acylethanolamine-hydrolyzing enzyme which is highly similar to acid ceramidase. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: FNVSLDSVPELRWLPVLRHYDLDLVRAAMAQVIGDRVPKWVHVLIGKVVLELERFLPQPFTGEIRGMCD
Specifications

Specifications

NP_055250
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FNVSLDSVPELRWLPVLRHYDLDLVRAAMAQVIGDRVPKWVHVLIGKVVLELERFLPQPFTGEIRGMCD
RUO
NAAA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27163
NAAA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASAHL/PLT
NAAA
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions