missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse STAT5b Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF1584
This item is not returnable.
View return policy
Description
STAT5b Polyclonal specifically detects STAT5b in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Specifications
| STAT5b | |
| Polyclonal | |
| Unconjugated | |
| signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6777 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Flow Cytometry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 1-5 ug/500 ug cell lysate, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, Knockout Validated | |
| STAT5B | |
| Human STAT5b synthetic peptide aa 777-787 | |
| 50 μg | |
| Primary | |
| Detects human and mouse STAT5b. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction