missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Myoglobin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18724467
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724467 25 μg 25µg
18780707 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18724467 Supplier R&D Systems Supplier No. MAB97203SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Myoglobin Monoclonal specifically detects Myoglobin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myoglobin
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 2269B
Conjugate Unconjugated
Dilution Western Blot 0.5 μg;/mL, Simple Western 10 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02144
Gene Alias MB, MGC13548, myoglobin, PVALB
Host Species Rabbit
Immunogen Purifed human cardiac Myoglobin antigen from human heart, Accession No. P02144
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4151.0
Test Specificity Detects human Myoglobin in direct ELISAs. Detects human, mouse, and rat Myoglabin in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.