missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Glutathione Peroxidase 4/GPX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB5457
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 4/GPX4 Monoclonal specifically detects Glutathione Peroxidase 4/GPX4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Glutathione Peroxidase 4/GPX4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P36969 | |
| GPX4 | |
| E. coli-derived recombinant human Glutathione Peroxidase 4 Gly74-Phe168 Accession # P36969 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat GPX4 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 565320 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GPX4, GSHPx-4, MCSP, PHGPx, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snGPx, snPHGPx, sperm nucleus glutathione peroxidase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2879 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction