missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Acetyl Histone H4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5215-SP
This item is not returnable.
View return policy
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Histone H4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| H4, H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4M, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, HIST4H4, histone 4, H4, Histone Cluster 1 H4, Histone Cluster 1 H4i, Histone Cluster 4 H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 121504 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P02310 | |
| HIST4H4 | |
| Tetrahymena pyriformis acetylated Histone H4 synthetic peptide AGGKGGKGMGKVGAKRHSK Accession # P02310 | |
| 25 μg | |
| Primary | |
| Detects immunogen in direct ELISAs and human Acetyl Histone H4 in Western blots. Reactivity with non-acetylated peptide was removed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction