Learn More
Invitrogen™ Human MLH3 (aa 1341-1426) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66338 (PA5-66338. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Specifications
| Accession Number | Q9UHC1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 27030 |
| Name | Human MLH3 (aa 1341-1426) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AV125803; BB126472; CAE; CAE1; cell surface glycoprotein; connexin 50; connexin-50; CTRCT1; C x 50; CZP1; DNA mismatch repair protein Mlh3; gap junction alpha 8; gap junction alpha-8 protein; gap junction membrane channel protein alpha-8; gap junction protein alpha 8; gap junction protein alpha 8 50 kDa; GJA8; HNPCC7; lens fiber protein MP70; lens intrinsic membrane protein MP70; MGC138372; MLH3; MP70; mutL homolog 3; mutL homolog 3 (E coli); MutL protein homolog 3 |
| Common Name | MLH3 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.