missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIBCO™ Human Microsomes, 20 Donors, Male Donors
Specifications
Specifications
| Donor Source | Pooled |
| Gender | Male |
| Physical Form | Frozen |
| Tissue | Liver |
| Host Species | Human |
| Concentration | 20 mg/mL |
| Content And Storage | Store between -60°C and -85°C. |
| Life Stage | Adult |
| Format | Tube |
| Sub Cellular Localization | Microsomes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?