missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC34821 Full-length ORF (NP_775857.1, 1 a.a. - 322 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_775857.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 283238 |
Molecular Weight (g/mol) | 62.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139843
|
Abnova™
H00283238-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129843
|
Abnova™
H00283238-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SLC22A24 belongs to a large family of transmembrane proteins that function as uniporters, symporters, and antiporters to transport organic ions across cell membranes (Jacobsson et al., 2007 [PubMed 17714910]).[supplied by OMIM]
Sequence: MGFDVLLDQVGGMGRFQICLIAFFCITNILLFPNIVLENFTAFTPSHRCWVPLLDNDSVSDNDTGTLSKDDLLRISIPLDSNLRPQKCQRFIHPQWQLLHLNGTFPNTNEPDTEPCVDGWVYDRSSFLSTIVTEWDLVCESQSLKSMVQSLFMAGSLLGGLIYGHLSDRVGRKIICKLCFLQLAISNTCAAFAPTFLVYCILRFLAGFSTMTILGNTFILSLEWTLPRSRSMTIMVLLCSYSVGQMLLGGLAFAIQDWHILQLTVSTPIIVLFLSSWYEQSPHSLPVSEAMVDIERKIVTPGICSVSGLVLSHDVHSTYCVTSpecifications
NP_775857.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGFDVLLDQVGGMGRFQICLIAFFCITNILLFPNIVLENFTAFTPSHRCWVPLLDNDSVSDNDTGTLSKDDLLRISIPLDSNLRPQKCQRFIHPQWQLLHLNGTFPNTNEPDTEPCVDGWVYDRSSFLSTIVTEWDLVCESQSLKSMVQSLFMAGSLLGGLIYGHLSDRVGRKIICKLCFLQLAISNTCAAFAPTFLVYCILRFLAGFSTMTILGNTFILSLEWTLPRSRSMTIMVLLCSYSVGQMLLGGLAFAIQDWHILQLTVSTPIIVLFLSSWYEQSPHSLPVSEAMVDIERKIVTPGICSVSGLVLSHDVHSTYCVT | |
RUO | |
SLC22A24 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
283238 | |
MGC34821 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC34821 | |
SLC22A24 | |
Recombinant | |
wheat germ expression system |