Learn More
STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array
Product Code. p-7217807
Encompass_Preferred
Click to view available options
Format:
96-well Plate Type 1
96-well Plate Type 3
Unit Size:
1 set
Product Code. 17179291
Brand: STEMCELL Technologies 07541
Please to purchase this item. Need a web account? Register with us today!
For characterization of human MSCs and their differentiation to chondrogenic, adipogenic, or osteogenic lineages
  • The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
  • MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
  • This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation
Specifications
Description Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts
Format 96-well Plate Type 1
For Use With (Application) Real Time PCR (qPCR)