missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAP3K6 Partial ORF (AAH15914, 225 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH15914 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9064 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139855
|
Abnova™
H00009064-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149855
|
Abnova™
H00009064-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the serine/threonine protein kinase family. The encoded kinase was identified by its interaction with MAP3K5/ASK, a protein kinase and an activator of c-Jun kinase (MAPK7/JNK) and MAPK14/p38 kinase. This kinase was found to weakly activate MAPK7, but not MAPK1/ERK or MAPK14. An alternatively spliced transcript variant has been found for this gene, but its biological validity has not been determined. [provided by RefSeq]
Sequence: HFWLHFLLQSCQPFKTACAQGDQCLVLVLEMNKVLLPAKLEVRGTDPVSTVTLSLLEPETQDIPSSWTFPVASICGVSASKRDERCCFLYALPPAQDVQLCSpecifications
AAH15914 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASK2/MAPKKK6/MGC125653/MGC20114 | |
MAP3K6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9064 | |
MAP3K6 (Human) Recombinant Protein (Q01) | |
HFWLHFLLQSCQPFKTACAQGDQCLVLVLEMNKVLLPAKLEVRGTDPVSTVTLSLLEPETQDIPSSWTFPVASICGVSASKRDERCCFLYALPPAQDVQLC | |
RUO | |
MAP3K6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |