Learn More
Abnova™ Human MANEA Full-length ORF (NP_078917.2, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079694-P01.10ug
Additional Details : Weight : 0.00010kg
Description
N-glycosylation of proteins is initiated in the endoplasmic reticulum (ER) by the transfer of the preassembled oligosaccharide glucose-3-mannose-9-N-acetylglucosamine-2 from dolichyl pyrophosphate to acceptor sites on the target protein by an oligosaccharyltransferase complex. This core oligosaccharide is sequentially processed by several ER glycosidases and by an endomannosidase (E.C. 3.2.1.130), such as MANEA, in the Golgi. MANEA catalyzes the release of mono-, di-, and triglucosylmannose oligosaccharides by cleaving the alpha-1,2-mannosidic bond that links them to high-mannose glycans (Hamilton et al., 2005 [PubMed 15677381]).[supplied by OMIM]
Sequence: MAKFRRRTCIILALFILFIFSLMMGLKMLRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRDPSVIETHMRQMRSASIGVLALSWYPPDVNDENGEPTDNLVPTILDKAHKYNLKVTFHIEPYSNRDDQNMYKNVKYIIDKYGNHPAFYRYKTKTGNALPMFYVYDSYITKPEKWANLLTTSGSRSIRNSPYDGLFIALLVEEKHKYDILQSGFDGIYTYFATNGFTYGSSHQNWASLKLFCDKYNLIFIPSVGPGYIDTSIRPWNTQNTRNRINGKYYEIGLSAALQTRPSLISITSFNEWHEGTQIEKAVPKRTSNTVYLDYRPHKPGLYLELTRKWSEKYSKERATYALDRQLPVSSpecifications
NP_078917.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686D20120/ENDO/FLJ12838/hEndo | |
MANEA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79694 | |
MANEA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKFRRRTCIILALFILFIFSLMMGLKMLRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRDPSVIETHMRQMRSASIGVLALSWYPPDVNDENGEPTDNLVPTILDKAHKYNLKVTFHIEPYSNRDDQNMYKNVKYIIDKYGNHPAFYRYKTKTGNALPMFYVYDSYITKPEKWANLLTTSGSRSIRNSPYDGLFIALLVEEKHKYDILQSGFDGIYTYFATNGFTYGSSHQNWASLKLFCDKYNLIFIPSVGPGYIDTSIRPWNTQNTRNRINGKYYEIGLSAALQTRPSLISITSFNEWHEGTQIEKAVPKRTSNTVYLDYRPHKPGLYLELTRKWSEKYSKERATYALDRQLPVS | |
RUO | |
MANEA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |