missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGEC1 Partial ORF (NP_005453.2, 848 a.a. - 918 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_005453.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9947 |
Molecular Weight (g/mol) | 33.44kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085185
|
Abnova™
H00009947-Q03.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075185
|
Abnova™
H00009947-Q03.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the melanoma antigen gene (MAGE) family. The proteins of this family are tumor-specific antigens that can be recognized by autologous cytolytic T lymphocytes. This protein contains a large number of unique short repetitive sequences in front of the MAGE-homologous sequence, and therefore is about 800 aa longer than the other MAGE proteins. [provided by RefSeq]
Sequence: ESPLQSPVISFSSSTSLSPFSEESSSPVDEYTSSSDTLLESDSLTDSESLIESEPLFTYTLDEKVDELARFSpecifications
NP_005453.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.44kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CT7/MAGE-C1/MGC39366 | |
MAGEC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9947 | |
MAGEC1 (Human) Recombinant Protein (Q03) | |
ESPLQSPVISFSSSTSLSPFSEESSSPVDEYTSSSDTLLESDSLTDSESLIESEPLFTYTLDEKVDELARF | |
RUO | |
MAGEC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |