missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-CSF Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2161
This item is not returnable.
View return policy
Description
M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| M-CSF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSF1 | |
| E. coli-derived recombinant human M-CSF | |
| 500 μg | |
| Primary | |
| Detects human M-CSF. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 26786 | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1435 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction