missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysosomal alpha-Glucosidase Antibody, R&D Systems™

Rabbit Monoclonal Antibody
208.00 € - 493.00 €
Specifications
| Antigen | LYAG/GAA |
|---|---|
| Clone | 2489C |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791787
![]() |
R&D Systems
MAB8329-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799996
![]() |
R&D Systems
MAB8329-100 |
100 μg |
493.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYAG/GAA Monoclonal specifically detects LYAG/GAA in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| LYAG/GAA | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Acid alpha-Glucosidase, Acid maltase, Aglucosidase alfa, EC 3.2.1.20, GAA, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase, Lysosomal alphaGlucosidase | |
| Human embryonic kidney cell, HEK293-derived human Lysosomal alpha-Glucosidase protein, Ala70-Cys952, Accession # P10253 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Lysosomal alpha-Glucosidase protein in direct ELISAs. |
| 2489C | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P10253 | |
| 2548 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title