Learn More
Invitrogen™ Human LXR alpha (aa 432-442) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP115
Additional Details : CAS : 69-74-9 Weight : 0.01000kg
Description
Human LXR Alpha (residues 432-442) Synthetic Peptide, PEP-115, from Invitrogen.
PEP-115 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 432-445 of human LXR alpha with an amino-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C-L432-R-L-Q-D-K-K-L-P-P-L-L-S-E445This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human LXR alpha and beta, catalog # PA1-331B. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
10062 | |
purified | |
Human LXR alpha (aa 432-442) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q13133 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
NR1H3 | |
No preservative | |
50 μg | |
C-L432-R-L-Q-D-K-K-L-P-P-L-L-S-E445 |