missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LSAMP Full-length ORF (AAH33803, 29 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH33803 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4045 |
Molecular Weight (g/mol) | 59.62kDa |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16137461
|
Abnova™
H00004045-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16127461
|
Abnova™
H00004045-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system. During development of the limbic system, this encoded protein is found on the surface of axonal membranes and growth cones, where it acts as a selective homophilic adhesion molecule, and guides the development of specific patterns of neuronal connections. [provided by RefSeq]
Sequence: VRSVDFNRGTDNITVRQGDTAILRCVVEDKNSKVAWLNRSGIIFAGHDKWSLDPRVELEKRHSLEYSLRIQKVDVYDEGSYTCSVQTQHEPKTSQVYLIVQVPPKISNISSDVTVNEGSNVTLVCMANGRPEPVITWRHLTPTGREFEGEEEYLEILGITREQSGKYECKAANEVSSADVKQVKVTVNYPPTITESKSNEATTGRQASLKCEASAVPAPDFEWYRDDTRINSANGLEIKSTEGQSSLTVTNVTEEHYGNYTCVAANKLGVTNASLVLFRPGSVRGINGSISLAVPLWLLAASLLCLLSKCSpécification
AAH33803 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.62kDa | |
Glutathione Sepharose 4 Fast Flow | |
VRSVDFNRGTDNITVRQGDTAILRCVVEDKNSKVAWLNRSGIIFAGHDKWSLDPRVELEKRHSLEYSLRIQKVDVYDEGSYTCSVQTQHEPKTSQVYLIVQVPPKISNISSDVTVNEGSNVTLVCMANGRPEPVITWRHLTPTGREFEGEEEYLEILGITREQSGKYECKAANEVSSADVKQVKVTVNYPPTITESKSNEATTGRQASLKCEASAVPAPDFEWYRDDTRINSANGLEIKSTEGQSSLTVTNVTEEHYGNYTCVAANKLGVTNASLVLFRPGSVRGINGSISLAVPLWLLAASLLCLLSKC | |
RUO | |
LSAMP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4045 | |
LSAMP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGLON3/LAMP | |
LSAMP | |
Recombinant | |
wheat germ expression system |