missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRP1B (aa 4526-4589) Control Fragment Recombinant Protein

Product Code. 30207941
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207941 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30207941 Supplier Invitrogen™ Supplier No. RP103985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64396 (PA5-64396. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the low density lipoprotein (LDL) receptor family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands. Disruption of this gene has been reported in several types of cancer. [provided by RefSeq, Jun 2016].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZR2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 53353
Name Human LRP1B (aa 4526-4589) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9630004P12Rik; EC 1.1.1.94; EC 3.4.21.9; LDL receptor related protein 1 B; low density lipoprotein receptor related protein-deleted in tumor; low density lipoprotein receptor-related protein 1 B; low density lipoprotein-related protein 1 B (deleted in tumors); low-density lipoprotein receptor-related protein 1 B; Low-density lipoprotein receptor-related protein-deleted in tumor; LRP1B; LRP-1 B; LRP-deleted in tumors; LRPDIT; LRP-DIT
Common Name LRP1B
Gene Symbol LRP1B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GGPSAFKLPHTAPPIYLNSDLKGPLTAGPTNYSNPVYAKLYMDGQNCRNSLGSVDERKELLPKK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.