missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA6/CD85b Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8656
This item is not returnable.
View return policy
Description
ILT8/CD85b/LILRA6 Monoclonal specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| ILT8/CD85b/LILRA6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LILRA6 | |
| HEK293 human embryonic kidney cell line-derived recombinant human LILRA6/CD85b Met1-Asn447 Accession # Q6PI73 | |
| 100 μg | |
| Primary | |
| Detects human LILRA6/CD85b in direct ELISA. Detects HEK293 cells transfected with LILRA6/CD85b but not irrelevant transfectants. | |
| Human | |
| Supernatant |
| Flow Cytometry, CyTOF | |
| 921330 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q6PI73 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79168 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu