missing translation for 'onlineSavingsMsg'
Learn More

Human LILRA3/CD85e Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18735527
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735527 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735527 Supplier R&D Systems Supplier No. AF2574SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

ILT6/CD85e/LILRA3 Polyclonal specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT6/CD85e/LILRA3
Applications Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8N6C8
Gene Alias CD85 antigen-like family member E, CD85e, CD85e antigen, HM31, HM43, ILT6, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31
Gene Symbols LILRA3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA3/CD85e Arg18-Glu439 Accession # Q8N6C8
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Test Specificity Detects human LILRA3/CD85e in direct ELISAs. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) ILT2, rhILT4, ILT5 is observed, and less than 5% cross-reactivity with rhLIR5 and rhLIR7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.