Learn More
Abnova™ Human LDOC1 Full-length ORF (NP_036449.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023641-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers. [provided by RefSeq]
Sequence: MVDELVLLLHALLMRHRALSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRAFLDEMKQCFGWDDDEDDDDEEEEDDYSpecifications
NP_036449.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCUR1/Mar7/Mart7 | |
LDOC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23641 | |
LDOC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVDELVLLLHALLMRHRALSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRAFLDEMKQCFGWDDDEDDDDEEEEDDY | |
RUO | |
LDOC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |