missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LBP Antibody, R&D Systems™
Goat Polyclonal Antibody
593.00 €
Specifications
| Antigen | LBP |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
LBP Polyclonal specifically detects LBP in Human samples. It is validated for Western Blot.Specifications
| LBP | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233 | |
| LBP | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| AAD21962 | |
| 3929 | |
| Mouse myeloma cell line NS0-derived recombinant human LBP Ala26-Val481 Accession # AAD21962 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human LBP in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse LBP is observed and less than 1% cross-reactivity with recombinant human BPI is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title