Learn More
Abnova™ Human KNTC1 Partial ORF (NP_055523, 2100 a.a. - 2209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009735-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division. Experimental evidence indicated that the encoded protein functioned in a similar manner to that of the Drosophila rough deal protein. [provided by RefSeq]
Sequence: DPQVILKQLEEHMNTGQLAGFSHQIRSLILNNIINKKEFGILAKTKYFQMLKMHAMNTNNITELVNYLANDLSLDEASVLITEYSKHCGKPVPPDTAPCEILKMFLSGLSSpecifications
NP_055523 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DPQVILKQLEEHMNTGQLAGFSHQIRSLILNNIINKKEFGILAKTKYFQMLKMHAMNTNNITELVNYLANDLSLDEASVLITEYSKHCGKPVPPDTAPCEILKMFLSGLS | |
RUO | |
KNTC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9735 | |
KNTC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ36151/KIAA0166/ROD | |
KNTC1 | |
Recombinant | |
wheat germ expression system |