Learn More
R&D Systems™ Human KLHDC2 (NP_055130.1) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Specifications
Specifications
| Gene | KLHDC2 |
| Accession Number | NP_055130.1 |
| Gene ID (Entrez) | 23588 |
| Gene Alias | HCA33, HCLP-1 Host cell factor-like protein 1, Hepatocellular carcinoma-associated antigen 33, Host cell factor homolog LCP, kelch domain containing 2, kelch domain-containing protein 2, LCP |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |