missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KLF15 Partial ORF (NP_054798.1, 1 a.a. - 78 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00028999-Q01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MVDHLLPVDENFSSPKCPVGYLGDRLVGRRAYHMLPSPVSEDDSDASSPCSCSSPDSQALCSCYGGGLGTESQDSILDSpezifikation
NP_054798.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.32kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVDHLLPVDENFSSPKCPVGYLGDRLVGRRAYHMLPSPVSEDDSDASSPCSCSSPDSQALCSCYGGGLGTESQDSILD | |
RUO | |
KLF15 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
28999 | |
KLF15 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779M1320|KKLF | |
KLF15 | |
Yes |