missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLF15 Full-length ORF (NP_054798.1, 1 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00028999-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138942

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Sequence: MVDHLLPVDENFSSPKCPVGYLGDRLVGRRAYHMLPSPVSEDDSDASSPCSCSSPDSQALCSCYGGGLGTESQDSILDFLLSQATLGSGGGSGSSIGASSGPVAWGPWRRAAAPVKGEHFCLPEFPLGDPDDVPRPFQPTLEEIEEFLEENMEPGVKEVPEGNSKDLDACSQLSAGPHKSHLHPGSSGRERCSPPPGGASAGGAQGPGGGPTPDGPIPVLLQIQPVPVKQESGTGPASPGQAPENVKVAQLLVNIQGQTFALVPQVVPSSNLNLPSKFVRIAPVPIAAKPVGSGPLGPGPAGLLMGQKFPKNPAAELIKMHKCTFPGCSKMYTKSSHLKAHLRRHTGEKPFACTWPGCGWRFSRSDELSRHRRSHSGVKPYQCPVCEKKFARSDHLSKHIKVHRFPRSSRSVRSVN
Specifications

Specifications

NP_054798.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp779M1320/KKLF
KLF15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
28999
KLF15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVDHLLPVDENFSSPKCPVGYLGDRLVGRRAYHMLPSPVSEDDSDASSPCSCSSPDSQALCSCYGGGLGTESQDSILDFLLSQATLGSGGGSGSSIGASSGPVAWGPWRRAAAPVKGEHFCLPEFPLGDPDDVPRPFQPTLEEIEEFLEENMEPGVKEVPEGNSKDLDACSQLSAGPHKSHLHPGSSGRERCSPPPGGASAGGAQGPGGGPTPDGPIPVLLQIQPVPVKQESGTGPASPGQAPENVKVAQLLVNIQGQTFALVPQVVPSSNLNLPSKFVRIAPVPIAAKPVGSGPLGPGPAGLLMGQKFPKNPAAELIKMHKCTFPGCSKMYTKSSHLKAHLRRHTGEKPFACTWPGCGWRFSRSDELSRHRRSHSGVKPYQCPVCEKKFARSDHLSKHIKVHRFPRSSRSVRSVN
RUO
KLF15
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions