missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KLF11 (aa 271-398) Control Fragment Recombinant Protein

Product Code. 30211786
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30211786 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30211786 Supplier Invitrogen™ Supplier No. RP106839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111465 (PA5-111465. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a zinc finger transcription factor that binds to SP1-like sequences in epsilon- and gamma-globin gene promoters. This binding inhibits cell growth and causes apoptosis. Defects in this gene are a cause of maturity-onset diabetes of the young type 7 (MODY7). Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14901
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8462
Name Human KLF11 (aa 271-398) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9830142A17; D12Ertd427e; FKLF; FKLF1; klf11; Krueppel-like factor 11; Kruppel like factor 11; Kruppel-like factor 11; MODY7; Tcfcp2l2; TGFB inducible early growth response 2; TGFB inducible early growth response 3; TGF-beta inducible early growth response protein 2; TGFB-inducible early growth response protein 2; TGFB-inducible early growth response protein 2 b; TGFB-inducible early growth response protein 3; Tieg2; TIEG-2; Tieg2b; Tieg3; TIEG-3; transcription factor CP2-like 2; transforming growth factor-beta-inducible early growth response protein 2; Transforming growth factor-beta-inducible early growth response protein 3
Common Name KLF11
Gene Symbol KLF11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KTTPLISVSVPAPPVLCQMIPVTGQSSMLPAFLKPPPQLSVGTVRPILAQAAPAPQPVFVGPAVPQGAVMLVLPQGALPPPAPCAANVMAAGNTKLLPLAPAPVFITSSQNCVPQVDFSRRRNYVCSF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.