Learn More
Proteintech™ Human JNK ELISA Kit
Product Code. 16861486 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species Unit Size
16861486 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16861486 Supplier Proteintech™ Supplier No. KE0007296T
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Solid phase sandwich ELISA for the quantitative determination of JNK in samples of human and mouse origin
  • For testing cell lysates
  • JNK (MAPK8, Mitogen-activated protein kinase 8, PRKM8, SAPK1, SAPK1C) belongs to MAP kinase subfamily
    • Activated by dual phosphorylation at Thr-Pro-Tyr motif during response to UV light
    • Functions as phosphorylate c-Jun at N-terminal serine regulatory sites of Ser-63 and Ser-73, mapping within transactivation domain
    • JNK activity abnormally elevated in obesity
    • Absence of JNK results in decreased adiposity, significantly improved insulin sensitivity and enhanced insulin receptor signaling capacity in two models of mouse obesity (ob/ob)
    • Mediator of obesity and insulin resistance and a potential target for therapeutics
Process:
  • Antibody specific for JNK pre-coated onto microwells
  • JNK protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody specific for JNK is added to detect captured JNK protein
  • For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Accession Number NP_001265476
Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.1 ng/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 5599
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P45983
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human JNK ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine