missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IRS2 Partial ORF (NP_003740.2, 494 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003740.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8660 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148985
|
Abnova™
H00008660-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138985
|
Abnova™
H00008660-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the insulin receptor substrate 2, a cytoplasmic signaling molecule that mediates effects of insulin, insulin-like growth factor 1, and other cytokines by acting as a molecular adaptor between diverse receptor tyrosine kinases and downstream effectors. The product of this gene is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation, as well as by an interleukin 4 receptor-associated kinase in response to IL4 treatment. [provided by RefSeq]
Sequence: DPGFMSLDEYGSSPGDLRAFCSHRSNTPESIAETPPARDGGGGGEFYGYMTMDRPLSHCGRSYRRVSGDAAQDLDRGLRKRTYSLTTPARQRPVPQPSSASLDEYTLMRASpecifications
NP_003740.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IRS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8660 | |
IRS2 (Human) Recombinant Protein (Q01) | |
DPGFMSLDEYGSSPGDLRAFCSHRSNTPESIAETPPARDGGGGGEFYGYMTMDRPLSHCGRSYRRVSGDAAQDLDRGLRKRTYSLTTPARQRPVPQPSSASLDEYTLMRA | |
RUO | |
IRS2 | |
Recombinant | |
wheat germ expression system |