missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 3/CD61 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2266
This item is not returnable.
View return policy
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Integrin beta 3/CD61 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P05106 | |
| ITGB3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin beta 3/CD61 Gly27-Asp718 Accession # P05106 | |
| 100 μg | |
| Primary | |
| Detects human Integrin beta 3/CD61 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Integrin beta 1, beta 2, recombinant mouse Integrin beta 4, or beta 6 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 256809 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD61, CD61 antigen, GP3A, GP3Aplatelet glycoprotein IIIa, GPIIIA, GPIIIaintegrin beta-3, INGRB3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, ITGB3, Platelet membrane glycoprotein IIIa | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3690 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction