missing translation for 'onlineSavingsMsg'
Learn More

Human Integrin alpha V beta 5 Alexa Fluor 594 Antibody, R&D Systems™

Product Code. 18772189
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772189 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772189 Supplier R&D Systems Supplier No. FAB2528T100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha V beta 5 Monoclonal specifically detects Integrin alpha V beta 5 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha V beta 5
Applications Flow Cytometry
Classification Monoclonal
Clone P5H9
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD51, integrin subunit alpha V, ITGAV, ITGB5, MSK8, Vitronectin Receptor alpha, VNRA, VTNR
Gene Symbols ITGAV
Host Species Mouse
Immunogen HT1080 human fibrosarcoma cell line
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Test Specificity Detects human Integrin alpha V beta 5. Recognizes the human Integrin alpha V beta 5 heterodimer and does not recognize the alpha V subunit in association with any other beta subunits.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.