missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Insulin R/CD220 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1544
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| Insulin R/CD220 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3643 | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 243524 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| NP_001073285 | |
| INSR | |
| Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys944 Accession # NP_001073285 | |
| 100 μg | |
| Primary | |
| Detects human Insulin R/CD220 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction