missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-4R alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF-230-SP
This item is not returnable.
View return policy
Description
IL-4R alpha Polyclonal specifically detects IL-4R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| IL-4R alpha | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL4R | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-4 R alpha | |
| 25 μg | |
| Primary | |
| Detects human IL-4 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) IL-1 sRI, rhIL-1 sRII, rhIL-2 sR alpha , rhIL-2 sR beta , rhIL-2 sR gamma , rhIL-3 sR alpha , and rhIL-5 sR alpha is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3566 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction