missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-37/IL-1F7 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
1 set
Description
The Human IL-37/IL-1F7 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-37/IL-1F7.
Specifications
Specifications
| Assay Range | 31.20 - 2000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 27178 |
| Gene Symbol | IL37 |
| Host Species | Human |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | IL-37/IL-1F7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction