Learn More
Proteintech™ Human IL-21 ELISA Kit
Product Code. 16801456 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Human
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species Unit Size
16801456 Human 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16801456 Supplier Proteintech™ Supplier No. KE0002296T
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous IL21
  • Recognizes human IL21
  • IL21 (Interleukin 21) is a cytokine produced predominantly by cluster of differentiation 4 (CD4+) T-cells and natural killer T-cells
    • IL21 receptor (IL21R) is a class I cytokine receptor, requiring dimerization with indispensable common gamma chain subunit to bind IL21
    • IL21 plays role in innate and adaptive immune responses by inducing differentiation, proliferation and activity of multiple target cells (macrophages, natural killer cells, B cells, cytotoxic T cells)
    • Mediates apoptosis in B-CLL cells through up-regulation of BIM, enhances both direct and antibody-dependent cellular cytotoxicity in cells
Process:
  • Antibody specific for IL21 pre-coated onto microwells
  • IL21 protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody of biotinylated specific for IL21 is added to detect captured IL21 protein
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Accession Number NP_068575
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 2 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 59067
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID Q9HBE4
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human Angiogenin ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine