missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-2 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC103561A-100
This item is not returnable.
View return policy
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-2 | |
| Monoclonal | |
| APC | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| E. coli -derived human IL-2, Ala21-Thr153, Accession # P60568 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1019308 | |
| 10 μL/106 cells | |
| P60568 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3558 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction