missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17RC Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2269
This item is not returnable.
View return policy
Description
IL-17RC Polyclonal specifically detects IL-17RC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| IL-17RC | |
| Polyclonal | |
| Unconjugated | |
| NP_116121 | |
| IL17RC | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RC Leu21-Ala454 Accession # NP_116121 | |
| 100 μg | |
| Primary | |
| Detects human IL-17 RC in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse IL-17 RC and 5% cross-reactivity with recombinant human (rh) IL-17 R, rhIL-17 RD, and rhIL-17B R is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| FLJ95963, FLJ96005, IL-17 RC, IL-17 receptor C, IL-17 RL, IL17F receptor, IL17RC, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 84818 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction