missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17/IL-17A Antibody, R&D Systems™

Mouse Monoclonal Antibody
245.00 € - 2090.00 €
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Clone | 41802R |
| Dilution | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754639
![]() |
R&D Systems
MAB3171R-SP |
25 μg |
245.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755239
![]() |
R&D Systems
MAB3171R-100 |
100 μg |
524.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18188810
|
R&D Systems
MAB3171R-01M |
1 mg |
2030.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18138820
|
R&D Systems
MAB3171R-500 |
500 ug |
2090.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.Specifications
| IL-17/IL-17A | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q16552 | |
| 3605 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 41802R | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects human IL-17 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer and no cross-reactivity with rhIL-17F, recombinant mouse IL-17, and recombinant rat IL-17A is observed. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title