missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 R alpha 1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF152
This item is not returnable.
View return policy
Description
IL-13R alpha 1 Polyclonal specifically detects IL-13R alpha 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Specifications
| IL-13R alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| Q5JSL4 | |
| IL13RA1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 1 Ala27-Thr343 (Thr130Ile) Accession # Q5JSL4 | |
| 100 μg | |
| Primary | |
| Detects human IL-13 R alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinat human (rh) IL-13 R alpha 2, recombinant mouse IL-13 R alpha 1, rhIL-5 R alpha , rhIL-5 R beta , rhIL-4 R, and rhIL-9 R is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 10-30 ug/mL, CyTOF-ready | |
| bB128O4.2.1 (interleukin 13 receptor, alpha 1), Cancer/testis antigen 19, CD213A1, CD213a1 antigen, CT19, IL13 receptor alpha-1 chain, IL-13 receptor subunit alpha-1, IL13R, IL13R alpha 1, IL-13R subunit alpha-1, IL-13Ra, IL13RA, IL13RA1, IL-13RA1, IL-13R-alpha-1, interleukin 13 receptor, alpha 1, interleukin-13 receptor subunit alpha-1, NR4 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 3597 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction