missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-11 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Plate
Unit Size:
1 set
Description
The Human IL-11 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-11 in cell culture supernates (10 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 1 Plate |
| Sensitivity | 8 pg/mL |
| Packaging | 96-well Strip Plate |
| Synonym | Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11 Oprelvekin, interleukin 11, interleukin-11, Oprelvekin |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | ELISA |
| Product Type | Solid Phase Sandwich ELISA |
| Specificity | Natural and recombinant human IL-11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction