Learn More
STEMCELL Technologies Human IL-10 ELISA Kit
Click to view available options
Quantity:
2 Plates
Unit Size:
Pack of 2
Description
- The Human Interleukin 10 (IL-10) ELISA Kit is designed for the quantitative detection and measurement of human IL-10 in biological fluids such as serum, plasma, and cell culture supernatants
- IL-10 is primarily produced by dendritic cells, macrophages, and CD4+ T cells
- IL-10 is important for the function of regulatory T cells and it enhances B cell proliferation, immunoglobulin secretion, and MHC class II expression
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Specifications
| Description | Dendritic Cells; Macrophages; Monocytes; T Cells, CD4+ |
| For Use With (Application) | ELISA |
| Quantity | 2 Plates |