missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 beta/IL-1F2 (Research Grade Canakinumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems MAB10349-SP
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3553 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| Hu110 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| Human IL-1 beta/IL-1F2 | |
| 25 μg | |
| Primary | |
| Detects Human IL-1 beta/IL-1F2 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Canakinumab. This product is for research use only. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction