missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 beta/IL-1F2 (Research Grade Canakinumab Biosimilar) Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems FAB10349S-100UG
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3553 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu110 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| Human IL-1 beta/IL-1F2 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction