Learn More
Invitrogen™ Human IgG (Fc) Recombinant Rabbit Monoclonal Antibody (4Z8R8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551235
Description
Immunogen sequence: PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG K.
Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. The Fc is a region of the immunoglobulin G (IgG) antibody that is responsible for binding to various immune cells and molecules, such as Fc receptors and complement proteinsIt is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifications
| Human IgG (Fc) | |
| Recombinant Monoclonal | |
| 1.01 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01857 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3500 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4Z8R8 | |
| Unconjugated | |
| IGHG1 | |
| IGHG1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.