Learn More
R&D Systems™ Human HMGB1/HMG-1 (NP_002119.1) VersaClone cDNA
Specifications
Specifications
| Gene | HMGB1/HMG-1 |
| Accession Number | NP_002119.1 |
| Gene ID (Entrez) | 3146 |
| Gene Alias | Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |