missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HM74A/PUMA-G/GPR109A PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems FAB2760P
This item is not returnable.
View return policy
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| HM74A/PUMA-G/GPR109A/NIACR1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| GPR109A, HCA2, HCAR2, hydroxycarboxylic acid receptor 2, Niacr1, Nicotinic Acid Receptor, PUMAG | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 338442 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 245106 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q8TDS4 | |
| HCAR2 | |
| NS0 mouse myeloma cell line transfected with human HM74A/PUMA-G/GPR109A Met1-Pro363 Accession # Q8TDS4 | |
| 100 Tests | |
| Primary | |
| Detects human HM74A/PUMA-G/GPR109A. Stains human HM74A/PUMA-G/GPR109A-transfected cells but not irrelevant transfectants. Cross-reactivity with HM74 (GPR109B) was not tested. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction