missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HIP1R Full-length ORF (NP_003950.1, 1 a.a. - 1068 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003950.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9026 |
Molecular Weight (g/mol) | 145.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193192
|
Abnova™
H00009026-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183192
|
Abnova™
H00009026-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MNSIKNVPARVLSRRPGHSLEAEREQFDKTQAISISKAINTQEAPVKEKHARRIILGTHHEKGAFTFWSYAIGLPLPSSSILSWKFCHVLHKVLRDGHPNVLHDCQRYRSNIREIGDLWGHLHDRYGQLVNVYTKLLLTKISFHLKHPQFPAGLEVTDEVLEKAAGTDVNNIFQLTVEMFDYMDCELKLSESVFRQLNTAIAVSQMSSGQCRLAPLIQVIQDCSHLYHYTVKLLFKLHSCLPADTLQGHRDRFHEQFHSLRNFFRRASDMLYFKRLIQIPRLPEGPPNFLRASALAEHIKPVVVIPEEAPEDEEPENLIEISTGPPAGEPVVVADLFDQTFGPPNGSVKDDRDLQIESLKREVEMLRSELEKIKLEAQRYIAQLKSQVNALEGELEEQRKQKQKALVDNEQLRHELAQLRAAQLEGERSQGLREEAERKASATEARYNKLKEKHSELVHVHAELLRKNADTAKQLTVTQQSQEEVARVKEQLAFQVEQVKRESELKLEEKSDQLEKLKRELEAKAGELARAQEALSHTEQSKSELSSRLDTLSAEKDALSGAVRQREADLLAAQSLVRETEAALSREQQRSSQEQGELQGRLAERESQEQGLRQRLLDEQFAVLRGAAAEAAGILQDAVSKLDDPLHLRCTSSPDYLVSRAQEALDAVSTLEEGHAQYLTSLADASALVAALTRFSHLAADTIINGGATSHLAPTDPADRLIDTCRECGARALELMGQLQDQQALRHMQASLVRTPLQGILQLGQELKPKSLDVRQEELGAVVDKEMAATSAAIEDAVRRIEDMMNQARHASSGVKLEVNERILNSCTDLMKAIRLLVTTSTSLQKEIVESGRGAATQQEFYAKNSRWTEGLISASKAVGWGATQLVEAADKVVLHTGKYEELIVCSHEIAASTAQLVAASKVKANKHSPHLSRLQECSRTVNERAANVVASTKSGQEQIEDRDTMDFSGLSLIKLKKQEMETQVRVLELEKTLEAERMRLGELRKQHYVLAGASGSPGEEVAIRPSTAPRSVTTKKPPLAQKPSVAPRQDHQLDKKDGIYPAQLVNYSpecifications
NP_003950.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
145.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNSIKNVPARVLSRRPGHSLEAEREQFDKTQAISISKAINTQEAPVKEKHARRIILGTHHEKGAFTFWSYAIGLPLPSSSILSWKFCHVLHKVLRDGHPNVLHDCQRYRSNIREIGDLWGHLHDRYGQLVNVYTKLLLTKISFHLKHPQFPAGLEVTDEVLEKAAGTDVNNIFQLTVEMFDYMDCELKLSESVFRQLNTAIAVSQMSSGQCRLAPLIQVIQDCSHLYHYTVKLLFKLHSCLPADTLQGHRDRFHEQFHSLRNFFRRASDMLYFKRLIQIPRLPEGPPNFLRASALAEHIKPVVVIPEEAPEDEEPENLIEISTGPPAGEPVVVADLFDQTFGPPNGSVKDDRDLQIESLKREVEMLRSELEKIKLEAQRYIAQLKSQVNALEGELEEQRKQKQKALVDNEQLRHELAQLRAAQLEGERSQGLREEAERKASATEARYNKLKEKHSELVHVHAELLRKNADTAKQLTVTQQSQEEVARVKEQLAFQVEQVKRESELKLEEKSDQLEKLKRELEAKAGELARAQEALSHTEQSKSELSSRLDTLSAEKDALSGAVRQREADLLAAQSLVRETEAALSREQQRSSQEQGELQGRLAERESQEQGLRQRLLDEQFAVLRGAAAEAAGILQDAVSKLDDPLHLRCTSSPDYLVSRAQEALDAVSTLEEGHAQYLTSLADASALVAALTRFSHLAADTIINGGATSHLAPTDPADRLIDTCRECGARALELMGQLQDQQALRHMQASLVRTPLQGILQLGQELKPKSLDVRQEELGAVVDKEMAATSAAIEDAVRRIEDMMNQARHASSGVKLEVNERILNSCTDLMKAIRLLVTTSTSLQKEIVESGRGAATQQEFYAKNSRWTEGLISASKAVGWGATQLVEAADKVVLHTGKYEELIVCSHEIAASTAQLVAASKVKANKHSPHLSRLQECSRTVNERAANVVASTKSGQEQIEDRDTMDFSGLSLIKLKKQEMETQVRVLELEKTLEAERMR | |
RUO | |
HIP1R | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9026 | |
HIP1R (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14000/FLJ27022/HIP12/HIP3/ILWEQ/KIAA0655/MGC47513 | |
HIP1R | |
Recombinant | |
wheat germ expression system |