missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Hepassocin/HFREP-1 ELISA Kit
Description
Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene.
Specifications
Specifications
| Accession Number | Q08830 |
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.1 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2267 |
| Gene Symbol | FGL1 |
| Interassay CV | <10% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?